Learn More
Instech Laboratories™ Feeding tube, rodent oral gavage, stainless steel, 22ga x 25mm, straight, sterile
Brand: Instech Laboratories™ FTSS-22S-25
This item is not returnable.
View return policy
Description
These traditional reusable gavage needles feature stainless steel tubes and ball-tips. They are sold straight but are easily bent to your desired curve for optimal placement. Available in seven sizes for mice and rats.- Stainless steel feeding tubes, 22ga x 25mm
- For rodent oral gavage
- 1.2mm round bulb
- Bendable
- Supplied as sterile pouch of 6 tubes