accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 100-81-2

CH3NH3

CAS RN 100-81-2

IUPAC Name: (3-methylphenyl)methanaminium
Synonyms: (3-methylphenyl)methanaminium
Molecular Weight (g/mol): 122.19
Molecular Formula: C8H12N
InChi Key: RGXUCUWVGKLACF-UHFFFAOYSA-O
SMILES: CC1=CC=CC(C[NH3+])=C1
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.