accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10042-88-3

H2OH2OH2OH2OH2OH2OClClClTb

CAS RN 10042-88-3

IUPAC Name: terbium(3+) hexahydrate trichloride
Synonyms: terbium(III) chloride
Molecular Weight (g/mol): 373.37
Molecular Formula: Cl3H12O6Tb
InChi Key: ULJUVCOAZNLCJZ-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.