accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10045-45-1

H3CNOHN

CAS RN 10045-45-1

IUPAC Name: 1-ethyl-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-ethyl-2-benzimidazolinone
Molecular Weight (g/mol): 162.19
Molecular Formula: C9H10N2O
InChi Key: CXUCKELNYMZTRT-UHFFFAOYSA-N
SMILES: CCN1C(=O)NC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.