accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10057-27-9

OFFFFFF

CAS RN 10057-27-9

IUPAC Name: hexafluoropropan-2-one trihydrate
Synonyms: hexafluoroacetone trihydrate
Molecular Weight (g/mol): 220.07
Molecular Formula: C3H6F6O4
InChi Key: SNZAEUWCEHDROX-UHFFFAOYSA-N
SMILES: O.O.O.FC(F)(F)C(=O)C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.