accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1006-99-1

H3CSClN

CAS RN 1006-99-1

IUPAC Name: 5-chloro-2-methyl-1,3-benzothiazole
Synonyms: 5-chloro-2-methyl-1,3-benzothiazole
Molecular Weight (g/mol): 183.65
Molecular Formula: C8H6ClNS
InChi Key: XCALAYIRFYALSX-UHFFFAOYSA-N
SMILES: CC1=NC2=CC(Cl)=CC=C2S1
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.