accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10061-01-5

CAS RN 10061-01-5

IUPAC Name: (1Z)-1,3-dichloroprop-1-ene
Synonyms: telone II C3H4cl2 ccris 227 cis-DCP
Molecular Weight (g/mol): 110.97
Molecular Formula: C3H4Cl2
InChi Key: UOORRWUZONOOLO-UPHRSURJSA-N
SMILES: ClC\C=C/Cl
For Use With (Equipment) 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.