accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1008-76-0

OO

CAS RN 1008-76-0

IUPAC Name: 5-phenyloxolan-2-one
Synonyms: 5-phenyloxolan-2-one
Molecular Weight (g/mol): 162.19
Molecular Formula: C10H10O2
InChi Key: AEUULUMEYIPECD-UHFFFAOYNA-N
SMILES: O=C1CCC(O1)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.