accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-16-6

CH3ONH

CAS RN 101-16-6

IUPAC Name: 3-methoxy-N-phenylaniline
Synonyms: 3-methoxy-N-phenylbenzenamine
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: MKASXAGBWHIGCF-UHFFFAOYSA-N
SMILES: COC1=CC=CC(NC2=CC=CC=C2)=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.