accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-20-2

CAS RN 101-20-2

IUPAC Name: 3-(4-chlorophenyl)-1-(3,4-dichlorophenyl)urea
Synonyms: triclocarban cutisan TCC nobacter solubacter triclocarbanum 3,4,4'-trichlorodiphenylurea
Molecular Weight (g/mol): 315.58
Molecular Formula: C13H9Cl3N2O
InChi Key: ICUTUKXCWQYESQ-UHFFFAOYSA-N
SMILES: ClC1=CC=C(NC(=O)NC2=CC=C(Cl)C(Cl)=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.