man

Test

CAS RN 101-43-9

H3CCH2OO

CAS RN 101-43-9

IUPAC Name: cyclohexyl 2-methylprop-2-enoate
Synonyms: cyclohexyl methacrylate
Molecular Weight (g/mol): 168.24
Molecular Formula: C10H16O2
InChi Key: OIWOHHBRDFKZNC-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1CCCCC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results