accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 101-77-9

NH2H2N

CAS RN 101-77-9

IUPAC Name: 4-[(4-aminophenyl)methyl]aniline
Synonyms: 4,4'-diaminodiphenylmethane 4,4'-diaminodiphenylmethan jeffamine AP-20 sumicure M epikure ddm MDA α-(p-aminophenyl)-p-toluidine bis-p-aminofenylmethan dianilinomethane p,p'-diaminodiphenylmethane
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: YBRVSVVVWCFQMG-UHFFFAOYSA-N
SMILES: NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
Quantity 
  • (4)
  • (1)
For Use With (Equipment) 
  • (2)
Percent Purity 
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
1x1 image pixel for data collection