accessibility menu, dialog, popup

Test

CAS RN 1017778-76-5

CH3OClOClF

CAS RN 1017778-76-5

IUPAC Name: 3-chloro-4-ethoxy-5-fluorobenzoyl chloride
Synonyms: 3-chloro-4-ethoxy-5-fluorobenzoyl chloride
Molecular Weight (g/mol): 237.05
Molecular Formula: C9H7Cl2FO2
InChi Key: DVFYPWIHSIEUNN-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(C=C1Cl)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results