accessibility menu, dialog, popup

Test

CAS RN 101990-69-6

NClClOH

CAS RN 101990-69-6

IUPAC Name: (2,6-dichloropyridin-4-yl)methanol
Synonyms: (2,6-dichloropyridin-4-yl)methanol
Molecular Weight (g/mol): 178.01
Molecular Formula: C6H5Cl2NO
InChi Key: YDGJTFCKLFLWFM-UHFFFAOYSA-N
SMILES: OCC1=CC(Cl)=NC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results