accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102123-28-4

CH3OOSNNH2

CAS RN 102123-28-4

IUPAC Name: methyl 3-amino-4-cyanothiophene-2-carboxylate
Synonyms: methyl 3-amino-4-cyanothiophene-2-carboxylate
Molecular Weight (g/mol): 182.2
Molecular Formula: C7H6N2O2S
InChi Key: AEWVEROSMGRHRG-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(N)C(=CS1)C#N
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.