accessibility menu, dialog, popup

Test

CAS RN 102308-32-7

CH3H3CCH3OONOH3CCH3O

CAS RN 102308-32-7

IUPAC Name: tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Synonyms: tert-butyl (4S)-4-formyl-2,2-dimethyl-1,3-oxazolidine-3-carboxylate
Molecular Weight (g/mol): 229.28
Molecular Formula: C11H19NO4
InChi Key: PNJXYVJNOCLJLJ-MRVPVSSYSA-N
SMILES: CC(C)(C)OC(=O)N1[C@@H](COC1(C)C)C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results