accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102392-83-6

NHNNH2HI

CAS RN 102392-83-6

IUPAC Name: pyrrolidine-1-carboximidamide hydroiodide
Synonyms: pyrrolidine-1-carboximidamide hydroiodide
Molecular Weight (g/mol): 241.08
Molecular Formula: C5H12IN3
InChi Key: XPASGQSDLPDWNV-UHFFFAOYSA-N
SMILES: I.NC(=N)N1CCCC1
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.