accessibility menu, dialog, popup

Test

CAS RN 102561-47-7

CAS RN 102561-47-7

IUPAC Name: butyl 4-isocyanatobenzoate
Synonyms: butyl 4-isocyanatobenzoate
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: PVCGQSQNUXNTNA-UHFFFAOYSA-N
SMILES: CCCCOC(=O)C1=CC=C(C=C1)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results