CAS RN 10264-55-8
CAS RN 10264-55-8
IUPAC Name:
3-ethylcyclopentan-1-one
Synonyms:
3-ethylcyclopentan-1-one
Molecular Weight (g/mol):
112.17
Molecular Formula:
C7H12O
InChi Key:
XERALSLWOPMNRJ-UHFFFAOYNA-N
SMILES:
CCC1CCC(=O)C1
1
–
1
of
1
results
1
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals
CAS: 10264-55-8 Molecular Formula: C7H12O Molecular Weight (g/mol): 112.172 MDL Number: MFCD00045445 InChI Key: XERALSLWOPMNRJ-UHFFFAOYSA-N PubChem CID: 139121 IUPAC Name: 3-ethylcyclopentan-1-one SMILES: CCC1CCC(=O)C1