accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 102767-28-2

CH3NONH2O

CAS RN 102767-28-2

IUPAC Name: (2R)-2-(2-oxopyrrolidin-1-yl)butanamide
Synonyms: levitiracetam levetiracetamum
Molecular Weight (g/mol): 170.21
Molecular Formula: C8H14N2O2
InChi Key: HPHUVLMMVZITSG-ZCFIWIBFSA-N
SMILES: CC[C@@H](N1CCCC1=O)C(N)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.