accessibility menu, dialog, popup

Test

CAS RN 1027730-68-2

CH3SOOOH

CAS RN 1027730-68-2

IUPAC Name: 2-(3-methanesulfinylphenyl)acetic acid
Synonyms: (3-methanesulfinylphenyl)acetic acid
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: DXFCQIDRHQFCHE-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=CC(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results