accessibility menu, dialog, popup

Test

CAS RN 1028141-88-9

PNNNNOONH2FPFFFFFRu

CAS RN 1028141-88-9

IUPAC Name: λ²-ruthenium(2+) bis(2,2'-bipyridine) hexafluoro-λ⁵-phosphanuide triphenylphosphane 4-aminobutanoate
Synonyms: λ²-ruthenium(2+) bis(bipyridyl) triphenylphosphine 4-aminobutyrate hexafluorophosphate
Molecular Weight (g/mol): 922.81
Molecular Formula: C42H39F6N5O2P2Ru
InChi Key: SWFWLLYVSYURHZ-UHFFFAOYSA-M
SMILES: [Ru++].NCCCC([O-])=O.F[P-](F)(F)(F)(F)F.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results