CAS RN 10284-63-6
CAS RN 10284-63-6
IUPAC Name:
(1R,2S,4S,5S)-6-methoxycyclohexane-1,2,3,4,5-pentol
Synonyms:
methylinositol
Molecular Weight (g/mol):
194.18
Molecular Formula:
C7H14O6
InChi Key:
DSCFFEYYQKSRSV-FEPQRWDDSA-N
SMILES:
COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O
1
–
2
of
2
results
1
Thermo Scientific Chemicals D-Pinitol, 95%
CAS: 10284-63-6 Molecular Formula: C7H14O6 Molecular Weight (g/mol): 194.18 MDL Number: MFCD00216659 InChI Key: DSCFFEYYQKSRSV-FEPQRWDDSA-N Synonym: d-pinitol,pinitol,3-o-methyl-d-chiro-inositol,d-+-pinitol,inzitol,pinit,+-pinitol,methylinositol,sennitol,d-ononitol PubChem CID: 164619 IUPAC Name: (1S,2S,4S,5R)-6-methoxycyclohexane-1,2,3,4,5-pentol SMILES: COC1[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]1O
2
D-Pinitol, MedChemExpress
MedChemExpress D-pinitol (3-O-Methyl-D-chiro-inositol) is a natural compound presented in several plants, like Pinaceae and Leguminosae plants. D-pinitol exerts hypoglycemic activity and protective effects in the cardiovascular system. D-pinitol has antiviral and larvicidal activities.