accessibility menu, dialog, popup

Test

CAS RN 1029439-83-5

BOOH3CCH3H3CH3CFOHF

CAS RN 1029439-83-5

IUPAC Name: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 256.06
Molecular Formula: C12H15BF2O3
InChi Key: KTQYJNVSVKKLGQ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(F)=C(O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results