accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1029623-72-0

FSH2NF

CAS RN 1029623-72-0

IUPAC Name: 2,5-difluorobenzene-1-carbothioamide
Synonyms: 2,5-difluorobenzenecarbothioamide
Molecular Weight (g/mol): 173.18
Molecular Formula: C7H5F2NS
InChi Key: SUFZOIVUVDOCQQ-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(F)C=CC(F)=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.