CAS RN 103-47-9
CAS RN 103-47-9
Thermo Scientific Chemicals CHES, 99%
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N Synonym: 2-(Cyclohexylamino)ethanesulfonic acid PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O
Thermo Scientific Chemicals CHES, 98+%, for biochemistry
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N Synonym: 2-(Cyclohexylamino)ethanesulfonic acid PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O
Thermo Scientific Chemicals CHES, 0.5M buffer soln, pH 9.0
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O
Melford CHES
CHES, also known as 2-(N-Cyclohexylamino) ethanesulfonic acid, is a chemical compound that plays a significant role in life science research, particularly in molecular biology and protein research.
CHES, 0.5M buffer soln, pH 9.5, Thermo Scientific™
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O
CHES, 0.5M buffer soln, pH 8.5, Thermo Scientific™
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O
CHES, 0.5M buffer soln, pH 10.0, Thermo Scientific™
CAS: 103-47-9 Molecular Formula: C8H17NO3S Molecular Weight (g/mol): 207.288 MDL Number: MFCD00003835 InChI Key: MKWKNSIESPFAQN-UHFFFAOYSA-N PubChem CID: 66898 IUPAC Name: 2-(cyclohexylamino)ethanesulfonic acid SMILES: C1CCC(CC1)NCCS(=O)(=O)O