accessibility menu, dialog, popup

Test

CAS RN 1031843-22-7

CH3NNNHCH3

CAS RN 1031843-22-7

IUPAC Name: methyl[(5-methyl-1-phenyl-1H-pyrazol-4-yl)methyl]amine
Synonyms: methyl[(5-methyl-1-phenylpyrazol-4-yl)methyl]amine
Molecular Weight (g/mol): 201.27
Molecular Formula: C12H15N3
InChi Key: PYMKNYBFTGMBJN-UHFFFAOYSA-N
SMILES: CNCC1=C(C)N(N=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results