CAS RN 1032358-00-1
CAS RN 1032358-00-1
IUPAC Name:
3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms:
3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol):
219.09
Molecular Formula:
C12H18BNO2
InChi Key:
MMCGEOWPLNVYGO-UHFFFAOYSA-N
SMILES:
CC1=C(C=CN=C1)B1OC(C)(C)C(C)(C)O1
1
–
1
of
1
results