accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1032758-87-4

BOOH3CCH3H3CH3CNNH3COOCH3CH3CH3

CAS RN 1032758-87-4

IUPAC Name: tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-methyl-N-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 334.22
Molecular Formula: C17H27BN2O4
InChi Key: QGBJKHALDLKAGX-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
1x1 image pixel for data collection
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.