CAS RN 103440-52-4
CAS RN 103440-52-4
IUPAC Name:
methyl 2-iodo-5-methylbenzoate
Synonyms:
methyl 2-iodo-5-methylbenzoate
Molecular Weight (g/mol):
276.07
Molecular Formula:
C9H9IO2
InChi Key:
DHYMLHOXGMUIDZ-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(I)C=CC(C)=C1
1
–
2
of
2
results
2
Methyl 2-iodo-5-methylbenzoate, 95%, Thermo Scientific Chemicals
CAS: 103440-52-4 Molecular Formula: C9H9IO2 Molecular Weight (g/mol): 276.07 MDL Number: MFCD12198129 InChI Key: DHYMLHOXGMUIDZ-UHFFFAOYSA-N PubChem CID: 14263946 IUPAC Name: methyl 2-iodo-5-methylbenzoate SMILES: COC(=O)C1=C(I)C=CC(C)=C1