man

Test

CAS RN 10365-98-7

CH3OBOHHO

CAS RN 10365-98-7

IUPAC Name: (3-methoxyphenyl)boronic acid
Synonyms: 3-methoxyphenylboronic acid
Molecular Weight (g/mol): 151.96
Molecular Formula: C7H9BO3
InChi Key: NLLGFYPSWCMUIV-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results