accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 10387-40-3

KCH3SHO

CAS RN 10387-40-3

IUPAC Name: ethanethioic S-acid potassium
Synonyms: thioacetic acid, potassium salt
Molecular Weight (g/mol): 115.21
Molecular Formula: C2H4KOS
InChi Key: SDJHDRMYZQFJJO-UHFFFAOYSA-N
SMILES: [K].CC(S)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.