accessibility menu, dialog, popup

Test

CAS RN 104-15-4

CH3SOHOO

CAS RN 104-15-4

IUPAC Name: 4-methylbenzene-1-sulfonic acid
Synonyms: para-toluene sulfonate P-toluenesulfonic acid benzenesulfonic acid, methyl- toluen-4-sulfonsaeure kyselina p-toluenesulfonova manro ptsa 65 LS tosic acid methylbenzenesulfonic acid methyl-benzenesulfonic acid manro ptsa 65 H
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: JOXIMZWYDAKGHI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)