CAS RN 104-29-0
CAS RN 104-29-0
IUPAC Name:
3-(4-chlorophenoxy)propane-1,2-diol
Synonyms:
chlorphenesin maolate gechophen mycil C9H11ClO3 chlorphenesine demykon chlorphenesin preparation 2-chlorphenesin chlorphenesin (substance)
Molecular Weight (g/mol):
202.63
Molecular Formula:
C9H11ClO3
InChi Key:
MXOAEAUPQDYUQM-UHFFFAOYNA-N
SMILES:
OCC(O)COC1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
3-(4-Chlorophenoxy)-1,2-propanediol, 99%, Thermo Scientific Chemicals
CAS: 104-29-0 Molecular Formula: C9H11ClO3 Molecular Weight (g/mol): 202.634 MDL Number: MFCD00021990 InChI Key: MXOAEAUPQDYUQM-UHFFFAOYSA-N Synonym: chlorphenesin,3-4-chlorophenoxy-1,2-propanediol,3-4-chlorophenoxy propane-1,2-diol,chlorophenesin,chlorphenesinum,adermykon,gecophen,demykon,mycil,chlorphenesine PubChem CID: 7697 ChEBI: CHEBI:3642 IUPAC Name: 3-(4-chlorophenoxy)propane-1,2-diol SMILES: C1=CC(=CC=C1OCC(CO)O)Cl
Promotions are available