accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 104-38-1

HOOOOH

CAS RN 104-38-1

IUPAC Name: 2-[4-(2-hydroxyethoxy)phenoxy]ethan-1-ol
Synonyms: 2-[4-(2-hydroxyethoxy)phenoxy]ethanol
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: WTPYFJNYAMXZJG-UHFFFAOYSA-N
SMILES: OCCOC1=CC=C(OCCO)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.