man

Test

CAS RN 104-93-8

H3COCH3

CAS RN 104-93-8

IUPAC Name: 1-methoxy-4-methylbenzene
Synonyms: P-methylanisole
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: CHLICZRVGGXEOD-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)