CAS RN 104439-77-2
CAS RN 104439-77-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-^m-hydrotetracarbonyldiruthenium(II), 98%, Thermo Scientific Chemicals
CAS: 104439-77-2 Molecular Formula: C62H42O6Ru2 MDL Number: MFCD03840556 Synonym: Shvo's Catalyst
Promotions are available