accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 169505-93-5

CAS RN 169505-93-5

IUPAC Name: [1-(5-chloro-1H-indol-3-yl)-2-methylpropan-2-yl]({2-[2-(cyclopropylmethoxy)phenoxy]ethyl})amine hydrochloride
Synonyms: [1-(5-chloro-1H-indol-3-yl)-2-methylpropan-2-yl]({2-[2-(cyclopropylmethoxy)phenoxy]ethyl})amine hydrochloride
Molecular Weight (g/mol): 449.42
Molecular Formula: C24H30Cl2N2O2
InChi Key: QFOPFGRPNPCPBX-UHFFFAOYSA-N
SMILES: Cl.CC(C)(CC1=CNC2=CC=C(Cl)C=C12)NCCOC1=CC=CC=C1OCC1CC1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.