CAS RN 23815-89-6
CAS RN 23815-89-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MedChemExpress Bradykinin (1-5)
MedChemExpress Bradykinin (1-5) is a major stable metabolite of Bradykinin, formed by the proteolytic action of angiotensin-converting enzyme (ACE).
2
MedChemExpress Bradykinin (1-5)
MedChemExpress Bradykinin (1-5) is a major stable metabolite of Bradykinin, formed by the proteolytic action of angiotensin-converting enzyme (ACE).