CAS RN 69004-03-1
CAS RN 69004-03-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Toltrazuril, MedChemExpress
MedChemExpress Toltrazuril (BAY-i 9142) is an antiprotozoal agent that acts upon Coccidia parasites.