accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 760207-87-2

CAS RN 760207-87-2

IUPAC Name: 5-bromo-2-methoxy-3-methylpyridine
Synonyms: 5-bromo-2-methoxy-3-methylpyridine
Molecular Weight (g/mol): 202.05
Molecular Formula: C7H8BrNO
InChi Key: RHYCFMXPPGZHAW-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.