accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 90600-20-7

CAS RN 90600-20-7

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}pent-4-enoate
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]pent-4-enoate
Molecular Weight (g/mol): 214.24
Molecular Formula: C10H16NO4
InChi Key: BUPDPLXLAKNJMI-ZETCQYMHSA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC=C)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.