accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 910036-84-9

CAS RN 910036-84-9

IUPAC Name: ({4-[(1H-imidazol-1-yl)methyl]phenyl}methyl)(methyl)amine; oxalic acid
Synonyms: oxalic acid; {[4-(imidazol-1-ylmethyl)phenyl]methyl}(methyl)amine
Molecular Weight (g/mol): 291.31
Molecular Formula: C14H17N3O4
InChi Key: AIQAEMDKVTYASZ-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CNCC1=CC=C(CN2C=CN=C2)C=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.