accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 912444-00-9

CAS RN 912444-00-9

IUPAC Name: 2-(2-methylpyrrolidin-2-yl)-1H-1,3-benzodiazole-4-carboxamide
Synonyms: 2-(2-methylpyrrolidin-2-yl)-1H-1,3-benzodiazole-4-carboxamide
Molecular Weight (g/mol): 244.3
Molecular Formula: C13H16N4O
InChi Key: JNAHVYVRKWKWKQ-UHFFFAOYNA-N
SMILES: CC1(CCCN1)C1=NC2=C(C=CC=C2N1)C(N)=O
Color 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Physical Form 
  • (1)
  • (5)
Grade 
  • (5)

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (5)
  • (1)
  • (5)
  • (1)
  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.