accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 92220-65-0

CAS RN 92220-65-0

IUPAC Name: iridium(3+) bis(2-(pyridin-2-yl)benzen-1-ide) chloride
Synonyms: iridium(3+) bis(2-(pyridin-2-yl)benzen-1-ide) chloride
Molecular Weight (g/mol): 536.05
Molecular Formula: C22H16ClIrN2
InChi Key: ORDROLZETYMBQO-UHFFFAOYSA-M
SMILES: [Cl-].[Ir+3].C1=CC=C([C-]=C1)C1=CC=CC=N1.C1=CC=C([C-]=C1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.