accessibility menu, dialog, popup

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 926921-61-1

CAS RN 926921-61-1

IUPAC Name: ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
Synonyms: ethyl 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylate
Molecular Weight (g/mol): 249.31
Molecular Formula: C13H19N3O2
InChi Key: MTWGDARCNWTWJN-UHFFFAOYNA-N
SMILES: CCOC(=O)C1CCCN(C1)C1=NC(C)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.