CAS RN 936500-94-6
CAS RN 936500-94-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Elinogrel, MedChemExpress
MedChemExpress Elinogrel (PRT060128) is a potent, direct acting, competitive, and reversible platelet P2Y12 antagonist (IC50=20 nM). It is orally and intravenously available and has potent antiplatelet effects.