CAS RN 948841-07-4
CAS RN 948841-07-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
TB5, MedChemExpress
MedChemExpress TB5 is a potent, selective and reversible inhibitor of hMAO-B with a Ki value of 0.11±0.01 μM.