missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HA Tag Horseradish Peroxidase-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems HAM0601
This item is not returnable.
View return policy
Description
HA Tag Monoclonal specifically detects HA Tag in Multi-Species samples. It is validated for Western Blot.
Specifications
| HA Tag | |
| Monoclonal | |
| HRP | |
| Supplied as a solution in PBS containing a preservative. with No Preservative | |
| HA, HA epitope tag, Hemagglutinin | |
| HA Tag Peptide Accession # ABB51961 | |
| 0.5 mL | |
| Primary | |
| Multi-species | |
| Purified |
| Western Blot | |
| 1049F | |
| Western Blot 1:1000-1:3000 dilution | |
| ABB51961 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects N-terminal and C-terminal HA-tagged proteins in Western blot. | |
| Do not freeze. 6 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction