missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Ethynylenedicyclohexanol, 97%, Thermo Scientific™

Product Code. 11414067
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-54-6
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00021400
InChI Key YMAIXWWPLZFJLA-UHFFFAOYSA-N
Synonym 1,1'-ethynylenedicyclohexanol, bis 1-hydroxycyclohexyl acetylene, 1-2-1-hydroxycyclohexyl ethynyl cyclohexan-1-ol, cyclohexanol, 1,1'-1,2-ethynediyl bis, cyclohexanol, 1,1'-ethynylenedi, cyclohexanol, 1,1'-1,2-ethynediyl bis-9ci, 1,1'-ethynylene-bis-cyclohexanol, 1-2-hydroxycyclohexyl ethynyl cyclohexan-1-ol, 1,1-ethynylenedicyclohexanol, 4-06-00-06076 beilstein handbook reference
PubChem CID 94825
IUPAC Name 1-[2-(1-hydroxycyclohexyl)ethynyl]cyclohexan-1-ol
SMILES OC1(CCCCC1)C#CC1(O)CCCCC1

Specifications

Quantity 100 g
Formula Weight 222.33
Percent Purity 97%
Chemical Name or Material 1,1'-Ethynylenedicyclohexanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'